missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 15499508
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000339-51-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD07368342
InChI Key DTGONJAUUOWYGB-UHFFFAOYSA-N
Synonym 3-fluoro-2-nitrobenzoic acid, 3-fluoro-2-nitrobenzoicacid, 2-carboxy-6-fluoronitrobenzene, pubchem4756, c7h4no4f, ksc498i1d, 3-fluoro-2-nitro-benzoic acid
PubChem CID 18915920
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 185.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-nitrobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'