missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-nitrophenol, 97%, Thermo Scientific Chemicals

Product Code. 15478228
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 385-01-3
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD07368753
InChI Key GAWNBKUTBVLIPL-UHFFFAOYSA-N
Synonym 3-fluoro-2-nitro-phenol, 2-nitro-3-fluorophenol, phenol, 3-fluoro-2-nitro, 3-fluoronitro-phenol, pubchem4124, acmc-209j0i, ksc497m1h
PubChem CID 2782777
IUPAC Name 3-fluoro-2-nitrophenol
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])O

Specifications

Quantity 1 g
Formula Weight 157.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.