missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-iodoaniline, 98%, Thermo Scientific Chemicals

Product Code. 15457968
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water (228.3 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 656-66-6
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD07774187
InChI Key KUVVJHBHRIXJKI-UHFFFAOYSA-N
Synonym 3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
PubChem CID 15475546
IUPAC Name 3-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1N)F)I

Specifications

Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Slightly soluble in water (228.3mg/L at 25°C).
Formula Weight 237.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-iodoaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.