missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methoxyphenylboronic acid, 97%, Thermo Scientific™
38.40 € - 141.40 €
Chemical Identifiers
CAS | 149507-26-6 |
---|---|
Molecular Formula | C7H8BFO3 |
Molecular Weight (g/mol) | 169.95 |
InChI Key | IILGLPAJXQMKGQ-UHFFFAOYSA-N |
Synonym | 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid |
PubChem CID | 2782194 |
IUPAC Name | (3-fluoro-4-methoxyphenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)OC)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
149507-26-6 | |
169.95 | |
3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid | |
(3-fluoro-4-methoxyphenyl)boronic acid |
C7H8BFO3 | |
IILGLPAJXQMKGQ-UHFFFAOYSA-N | |
2782194 | |
B(C1=CC(=C(C=C1)OC)F)(O)O |
Specifications
149507-26-6 | |
96% min. (HPLC) | |
C7H8BFO3 | |
Solubility in water: .. Other solubilities: soluble in methanol and thf | |
B(C1=CC(=C(C=C1)OC)F)(O)O | |
169.95 | |
169.95 | |
3-Fluoro-4-methoxyphenylboronic acid |
211.0°C to 213.0°C | |
Glass Bottle | |
3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid | |
IILGLPAJXQMKGQ-UHFFFAOYSA-N | |
(3-fluoro-4-methoxyphenyl)boronic acid | |
2782194 | |
97% |
Safety and Handling
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning