missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15409507
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609807-25-2
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD07363780
InChI Key MQFKLCWETPKBCH-UHFFFAOYSA-N
Synonym 3-fluoro-5-methoxybenzeneboronic acid, 3-fluoro-5-methoxyphenyl boronic acid, boronic acid, 3-fluoro-5-methoxyphenyl, 3-fluoro-5-methoxyphenylboronicacid, 3-fluoro-5-methoxy-phenyl boronic acid, acmc-1b9ik, 3-fluoro-5-methoxybenzeneboronicacid, 3-fluoro-5-methoxy phenylboronic acid, 3-fluoro-5-methoxyphenylboronic acid
PubChem CID 2763243
IUPAC Name (3-fluoro-5-methoxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)OC)(O)O

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-methoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'