missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoroazetidine hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 15415788

3-Fluoroazetidine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15415788
250 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15415788

Brand: Thermo Scientific Alfa Aesar H59454.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617718-46-4
Molecular Formula C3H7ClFN
Molecular Weight (g/mol) 111.544
MDL Number MFCD05663715
InChI Key PXFUWRWCKSLCLS-UHFFFAOYSA-N
Synonym 3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt
PubChem CID 10125054
IUPAC Name 3-fluoroazetidine;hydrochloride
SMILES C1C(CN1)F.Cl

Specifications

Quantity 250 mg
Formula Weight 111.55
Percent Purity 95%
Chemical Name or Material 3-Fluoroazetidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF29Y035 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberH59454.ME,H59454.MD
Lot NumberN06J031 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog NumberH59454.ME,H59454.MD
Lot NumberM21I038 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog NumberH59454.ME,H59454.MD
Lot NumberF29Y035 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59454.ME,H59454.MD
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoroazetidine hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.