missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetone, 98%, Thermo Scientific™

Product Code. 10008731
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1737-19-5
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00061144
InChI Key UWCPYXSRCQVABG-UHFFFAOYSA-N
Synonym 3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
PubChem CID 137185
IUPAC Name 1-(3-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4955 to 1.4975 (20°C, 589nm)
Quantity 1 g
Formula Weight 152.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenylacetone, 98%
Show More Show Less
Product Identifier
  • 3-Fluorophenylacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.