missing translation for 'onlineSavingsMsg'
Learn More
3-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals Product Code.: 11464993

3-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 11464993
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11464993

Brand: Thermo Scientific Alfa Aesar A12275.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-39-1
Molecular Formula C8H3FO3
Molecular Weight (g/mol) 166.11
MDL Number MFCD00039696
InChI Key WWJAZKZLSDRAIV-UHFFFAOYSA-N
Synonym 3-fluorophthalic anhydride, 4-fluoroisobenzofuran-1,3-dione, 3-fluorophthalicanhydride, 4-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 4-fluoro, 5-fluoro-isobenzofurandione, 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluor-2-benzofuran-1,3-dion, pubchem1949, fluorophthalic anhydride
PubChem CID 69551
IUPAC Name 4-fluoro-2-benzofuran-1,3-dione
SMILES FC1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Beilstein 1283580
Sensitivity Moisture sensitive
Formula Weight 166.11
Percent Purity 98%
Chemical Name or Material 3-Fluorophthalic anhydride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ05K016 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog NumberA12275.14,A12275.06
Lot NumberX04K036 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog NumberA12275.14,A12275.06
Lot Number10217192 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12275.14,A12275.06
Lot NumberG6184A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12275.14,A12275.06
Lot Number10111255 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12275.14,A12275.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.