missing translation for 'onlineSavingsMsg'
Learn More

3-Furoic acid, 99%

Product Code. 10489300 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10489300

Brand: Thermo Scientific Acros 172450250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-93-7 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-93-7
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005350
InChI Key IHCCAYCGZOLTEU-UHFFFAOYSA-N
Synonym 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee
PubChem CID 10268
ChEBI CHEBI:30846
IUPAC Name furan-3-carboxylic acid
SMILES OC(=O)C1=COC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121°C to 122°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 25 g
Beilstein 18, I, 439
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 112.08
Percent Purity 99%
Chemical Name or Material 3-Furoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Furoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.