missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one, 97%, Thermo Scientific Chemicals

Product Code. 11317395
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Specifications

Density 0.88
Boiling Point 67°C to 68°C (50 mmHg)
Flash Point 39°C (102°F)
Refractive Index 1.441
Quantity 10 g
UN Number UN1992
Beilstein 1699613
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 3-Hexyn-2-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'