missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexyne, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
Specifications
Specifications
Density | 0.721 |
Boiling Point | 80°C to 82°C |
Flash Point | −10°C (14°F) |
Odor | Characteristic |
Linear Formula | CH3CH2C≡CCH2CH3 |
Refractive Index | 1.411 |
Quantity | 5 g |
UN Number | UN3295 |
Beilstein | 1731158 |
Solubility Information | Not miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'