missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%, Thermo Scientific Chemicals

51.30 € - 421.00 €

Chemical Identifiers

CAS 53279-72-4
Molecular Formula C7H3I3O3
Molecular Weight (g/mol) 515.81
MDL Number MFCD00020305
InChI Key GIAVHGFPMPSIFI-UHFFFAOYSA-N
Synonym htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
PubChem CID 96627
IUPAC Name 3-hydroxy-2,4,6-triiodobenzoic acid
SMILES OC(=O)C1=C(I)C=C(I)C(O)=C1I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420967
View Documents
Thermo Scientific Alfa Aesar
B23355.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11430967
View Documents
Thermo Scientific Alfa Aesar
B23355.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11440967
View Documents
Thermo Scientific Alfa Aesar
B23355.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53279-72-4
515.81
GIAVHGFPMPSIFI-UHFFFAOYSA-N
96627
OC(=O)C1=C(I)C=C(I)C(O)=C1I
C7H3I3O3
MFCD00020305
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
3-hydroxy-2,4,6-triiodobenzoic acid

Specifications

53279-72-4
C7H3I3O3
2113494
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
OC(=O)C1=C(I)C=C(I)C(O)=C1I
515.81
515.81
3-Hydroxy-2,4,6-triiodobenzoic acid
211°C to 215°C
MFCD00020305
Light sensitive
GIAVHGFPMPSIFI-UHFFFAOYSA-N
3-hydroxy-2,4,6-triiodobenzoic acid
96627
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

missing translation for 'einecsNumber' : 258-457-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.