missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals

39.50 € - 134.67 €

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
Synonym maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
PubChem CID 8369
ChEBI CHEBI:69438
IUPAC Name 3-hydroxy-2-methylpyran-4-one
SMILES CC1=C(O)C(=O)C=CO1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11439612
View Documents
Thermo Scientific Alfa Aesar
A10735.14
25 g
39.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
11449612
View Documents
Thermo Scientific Alfa Aesar
A10735.22
100 g
134.67 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

118-71-8
126.11
XPCTZQVDEJYUGT-UHFFFAOYSA-N
8369
3-hydroxy-2-methylpyran-4-one
C6H6O3
MFCD00006578
maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
CHEBI:69438
CC1=C(O)C(=O)C=CO1

Specifications

118-71-8
170°C (100 mmHg)
C6H6O3
112169
maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
CC1=C(O)C(=O)C=CO1
126.11
CHEBI:69438
99%
161°C to 165°C
95°C (203°F)
MFCD00006578
14,5713
XPCTZQVDEJYUGT-UHFFFAOYSA-N
3-hydroxy-2-methylpyran-4-one
8369
126.11
3-Hydroxy-2-methyl-4-pyrone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

missing translation for 'einecsNumber' : 204-271-8

missing translation for 'rtecsNumber' : UQ1050000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only