missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11417054
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-30-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002511
InChI Key ZQLCWPXBHUALQC-UHFFFAOYSA-N
Synonym 3-hydroxy-p-toluic acid, 3-hydroxy-4-methyl-benzoic acid, 3,4-cresotic acid, benzoic acid, 3-hydroxy-4-methyl, 3-hydroxy-4-methylbenzoicacid, pubchem3890, acmc-209m5x, 3-hydroxy-4-methyl benzoic acid, 3,4-cresotic acid; 3-hydroxy-p-toluic acid, 3-hydroxy-4-methylbenzoic acid, technical grade
PubChem CID 68512
IUPAC Name 3-hydroxy-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1O)C(O)=O

Specifications

Melting Point 207°C to 212°C
Odor Odorless
Quantity 5 g
Beilstein 1936497
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'