missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 85.0°C to 90.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Argentometry) |
Quantity | 5 g |
Formula Weight | 109.56 |
Percent Purity | 97% |
Chemical Name or Material | 3-Hydroxyazetidine hydrochloride |
Safety and Handling

Product Identifier
- 3-HYDROXYAZETIDINE HYDROCHLORIDE, 97%
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction