missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.
Solubility
Soluble in DMSO and Methanol
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.
Solubility
Soluble in DMSO and Methanol
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 102°C to 105°C |
Boiling Point | 191°C (50 mmHg) |
Quantity | 500 g |
Beilstein | 507099 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in DMSO and Methanol |
Formula Weight | 122.12 |
Percent Purity | 97% |
Chemical Name or Material | 3-Hydroxybenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'