missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde, 98.5%, Thermo Scientific Chemicals

Product Code. 10636502
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-83-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003368
InChI Key IAVREABSGIHHMO-UHFFFAOYSA-N
Synonym m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
PubChem CID 101
ChEBI CHEBI:16207
IUPAC Name 3-hydroxybenzaldehyde
SMILES C1=CC(=CC(=C1)O)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 101°C to 105°C
Color Brown to Yellow
Boiling Point 191°C (50.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4CHO
Quantity 100 g
Beilstein 08, 58
Solubility Information Solubility in water: soluble in water
Formula Weight 122.12
Percent Purity 98.50%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzaldehyde, 98.5%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.