missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10774051
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 201.0°C to 204.0°C
Color White
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HOC6H4CO2H
Quantity 100 g
Beilstein 10, 134
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot alcohol and ether
Formula Weight 138.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Hydroxybenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'