Learn More
3-Hydroxybenzonitrile, 99%, Thermo Scientific Chemicals
38.29 € - 128.64 €
Chemical Identifiers
CAS | 873-62-1 |
---|---|
Molecular Formula | C7H5NO |
Molecular Weight (g/mol) | 119.123 |
MDL Number | MFCD00002252 |
InChI Key | SGHBRHKBCLLVCI-UHFFFAOYSA-N |
Synonym | 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 |
PubChem CID | 13394 |
IUPAC Name | 3-hydroxybenzonitrile |
SMILES | C1=CC(=CC(=C1)O)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430297
|
Thermo Scientific Alfa Aesar
B21702.06 |
5 g |
38.29 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440297
|
Thermo Scientific Alfa Aesar
B21702.14 |
25 g |
128.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
873-62-1 | |
119.123 | |
SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
13394 | |
C1=CC(=CC(=C1)O)C#N |
C7H5NO | |
MFCD00002252 | |
3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
3-hydroxybenzonitrile |
Specifications
873-62-1 | |
C7H5NO | |
3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
3-hydroxybenzonitrile | |
13394 | |
99% |
78°C to 81°C | |
MFCD00002252 | |
Slightly soluble in water. | |
C1=CC(=CC(=C1)O)C#N | |
119.123 | |
119.12 | |
3-Hydroxybenzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 212-845-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.