missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 10428850
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 241°C to 243°C
Color White
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2CH2NH2·HCl
Quantity 25 g
Beilstein 13, I, 325
Merck Index 15, 3470
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol and hot 95% ethanol,practically insoluble in ether,chloroform,,benzene and toluene
Formula Weight 189.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxytyramine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'