missing translation for 'onlineSavingsMsg'
Learn More
3-Iodo-1-phenylpropane, 97%, Thermo Scientific Chemicals Product Code.: 15470208

3-Iodo-1-phenylpropane, 97%, Thermo Scientific Chemicals

Product Code. 15470208
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15470208

Brand: Thermo Scientific Alfa Aesar H53420.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4119-41-9
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.09
MDL Number MFCD00019036
InChI Key RGCKJSPKMTWLLX-UHFFFAOYSA-N
Synonym 1-iodo-3-phenylpropane, 3-iodopropyl benzene, benzene, 3-iodopropyl, g-phenyl propyliodine, 3-phenylpropyl iodide, l-iodo-3-phenylpropane, acmc-20aou9, 3-phenyl-1-iodopropane, phch2ch2ch2i, 3-iodo-1-phenylpropane
PubChem CID 138121
IUPAC Name 3-iodopropylbenzene
SMILES ICCCC1=CC=CC=C1

Specifications

Density 1.53
Flash Point 110°C (230°F)
Linear Formula C6H5(CH2)3I
Refractive Index 1.582
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 97%
Chemical Name or Material 3-Iodo-1-phenylpropane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Iodo-1-phenylpropane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.