missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-n-propoxypyridine, 97%, Thermo Scientific™

114.00 € - 507.00 €

Chemical Identifiers

CAS 902837-45-0
Molecular Formula C8H10INO
Molecular Weight (g/mol) 263.078
MDL Number MFCD07781164
InChI Key WWMBXRKMRZDRFH-UHFFFAOYSA-N
Synonym 3-iodo-2-n-propoxypyridine, 3-iodo-2-propoxy-pyridine
PubChem CID 42553132
IUPAC Name 3-iodo-2-propoxypyridine
SMILES CCCOC1=C(C=CC=N1)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494530
View Documents
Thermo Scientific Alfa Aesar
H50101.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11404540
View Documents
Thermo Scientific Alfa Aesar
H50101.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

902837-45-0
263.078
WWMBXRKMRZDRFH-UHFFFAOYSA-N
42553132
CCCOC1=C(C=CC=N1)I
C8H10INO
MFCD07781164
3-iodo-2-n-propoxypyridine, 3-iodo-2-propoxy-pyridine
3-iodo-2-propoxypyridine

Specifications

902837-45-0
MFCD07781164
3-iodo-2-n-propoxypyridine, 3-iodo-2-propoxy-pyridine
CCCOC1=C(C=CC=N1)I
263.078
263.08
3-Iodo-2-n-propoxypyridine
C8H10INO
Light sensitive
WWMBXRKMRZDRFH-UHFFFAOYSA-N
3-iodo-2-propoxypyridine
42553132
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.