Learn More
3-Iodochromone, 97%, Thermo Scientific Chemicals
109.00 € - 294.00 €
Chemical Identifiers
CAS | 122775-34-2 |
---|---|
Molecular Formula | C9H5IO2 |
Molecular Weight (g/mol) | 272.04 |
MDL Number | MFCD09475514 |
InChI Key | SZDBDDKQJHHRGH-UHFFFAOYSA-N |
Synonym | 3-iodochromone, 3-iodo-4h-chromen-4-one, 3-iodo-4h-benzopyran-4-one |
PubChem CID | 10901720 |
IUPAC Name | 3-iodo-4H-chromen-4-one |
SMILES | IC1=COC2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15407237
|
Thermo Scientific Alfa Aesar
H34295.MD |
250 mg |
109.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15497227
|
Thermo Scientific Alfa Aesar
H34295.03 |
1 g |
294.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.
Notes
Light sensitive. Store in the dark. Store away from oxidizing agent, light.
Chemical Identifiers
122775-34-2 | |
272.04 | |
SZDBDDKQJHHRGH-UHFFFAOYSA-N | |
10901720 | |
IC1=COC2=CC=CC=C2C1=O |
C9H5IO2 | |
MFCD09475514 | |
3-iodochromone, 3-iodo-4h-chromen-4-one, 3-iodo-4h-benzopyran-4-one | |
3-iodo-4H-chromen-4-one |
Specifications
122775-34-2 | |
C9H5IO2 | |
Light sensitive | |
SZDBDDKQJHHRGH-UHFFFAOYSA-N | |
3-iodo-4H-chromen-4-one | |
10901720 | |
97% |
95°C to 97°C | |
MFCD09475514 | |
3-iodochromone, 3-iodo-4h-chromen-4-one, 3-iodo-4h-benzopyran-4-one | |
IC1=COC2=CC=CC=C2C1=O | |
272.04 | |
272.04 | |
3-Iodochromone |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only