missing translation for 'onlineSavingsMsg'
Learn More

3-Iodooxetane, 95%, Thermo Scientific Chemicals

Product Code. 15493808
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26272-85-5
Molecular Formula C3H5IO
Molecular Weight (g/mol) 183.98
MDL Number MFCD08544403
InChI Key KBEIFKMKVCDETC-UHFFFAOYSA-N
Synonym 3-iodo-oxetane, 1,3-epoxy-2-iodopropane, oxetane, 3-iodo, 3-iodanyloxetane, 3-iodooxetane, acmc-1cgai, 2-iodo-1,3-epoxypropane, 1,3-?epoxy-?2-?iodopropane
PubChem CID 13356686
IUPAC Name 3-iodooxetane
SMILES IC1COC1

Specifications

Density 2.14
Boiling Point 166°C
Flash Point 54°C (129°F)
Quantity 500 mg
UN Number UN1993
Sensitivity Light sensitive
Formula Weight 183.98
Percent Purity 95%
Chemical Name or Material 3-Iodooxetane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.