Learn More
(3-Isocyanatopropyl)triethoxysilane, 95%, Thermo Scientific Chemicals
27.20 € - 44.90 €
Chemical Identifiers
CAS | 24801-88-5 |
---|---|
Molecular Formula | C10H21NO4Si |
Molecular Weight (g/mol) | 247.366 |
MDL Number | MFCD00051459 |
InChI Key | FRGPKMWIYVTFIQ-UHFFFAOYSA-N |
Synonym | 3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes |
PubChem CID | 90613 |
IUPAC Name | triethoxy(3-isocyanatopropyl)silane |
SMILES | CCO[Si](CCCN=C=O)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475333
|
Thermo Scientific Alfa Aesar
A12890.06 |
5 g |
27.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485333
|
Thermo Scientific Alfa Aesar
A12890.14 |
25 g |
44.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
24801-88-5 | |
247.366 | |
FRGPKMWIYVTFIQ-UHFFFAOYSA-N | |
90613 | |
CCO[Si](CCCN=C=O)(OCC)OCC |
C10H21NO4Si | |
MFCD00051459 | |
3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes | |
triethoxy(3-isocyanatopropyl)silane |
Specifications
238°C | |
C10H21NO4Si | |
MFCD00051459 | |
4404652 | |
3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes | |
FRGPKMWIYVTFIQ-UHFFFAOYSA-N | |
triethoxy(3-isocyanatopropyl)silane | |
90613 | |
95% | |
1.421 | |
0.99 |
24801-88-5 | |
(CH3CH2O)3SiCH2CH2CH2NCO | |
UN2927 | |
77°C (170°F) | |
Hydrolyzes in water. | |
CCO[Si](CCCN=C=O)(OCC)OCC | |
247.366 | |
247.37 | |
Mild | |
Moisture sensitive | |
(3-Isocyanatopropyl)triethoxysilane |
Safety and Handling
GHS H Statement
H330-H334-H314-H318-H227-H302-H312
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P235-P260-P264b-P270-P271-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314-H330-H334
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 246-467-6
missing translation for 'rtecsNumber' : VV6691000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only