missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
50 mL
250 mL
1 L
Description
(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 96°C (30mmHg) |
Color | Colorless |
Physical Form | Liquid |
Quantity | 50 mL |
Linear Formula | HSCH2CH2CH2Si(OCH3)2CH3 |
Flash Point | 93°C (199°F) |
Solubility Information | Insoluble in water. |
Formula Weight | 180.34 |
Percent Purity | 95% |
Refractive Index | 1.45 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'