missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals

Product Code. 11334598
Click to view available options
Quantity:
10 mL
50 mL
250 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31001-77-1
Molecular Formula C6H16O2SSi
Molecular Weight (g/mol) 180.337
MDL Number MFCD00053923
InChI Key IKYAJDOSWUATPI-UHFFFAOYSA-N
Synonym 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan
PubChem CID 2759524
IUPAC Name 3-[dimethoxy(methyl)silyl]propane-1-thiol
SMILES CO[Si](C)(CCCS)OC

Specifications

Boiling Point 96°C (30mmHg)
Color Colorless
Physical Form Liquid
Quantity 1 L
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
Flash Point 93°C (199°F)
Solubility Information Insoluble in water.
Formula Weight 180.34
Percent Purity 95%
Refractive Index 1.45
Sensitivity Moisture sensitive
Density 1.000 g/mL
Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.