missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)triethoxysilane, 94%, Thermo Scientific Chemicals

Product Code. 11494687
Click to view available options
Quantity:
25 g
100 g

It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

Solubility
Hydrolyzes slowly in water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14814-09-6
Molecular Formula C9H22O3SSi
Molecular Weight (g/mol) 238.417
MDL Number MFCD00053924
InChI Key DCQBZYNUSLHVJC-UHFFFAOYSA-N
Synonym 3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy
PubChem CID 84658
IUPAC Name 3-triethoxysilylpropane-1-thiol
SMILES CCO[Si](CCCS)(OCC)OCC

Specifications

Boiling Point 210°C
Quantity 25 g
Linear Formula (CH3CH2O)3Si(CH2)3SH
Beilstein 2039575
Flash Point 88°C (190°F)
Solubility Information Hydrolyzes slowly in water.
Formula Weight 238.42
Percent Purity 94%
Odor Unpleasant
Refractive Index 1.437
Sensitivity Air and moisture sensitive
Density 0.98
Chemical Name or Material (3-Mercaptopropyl)triethoxysilane
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • (3-Mercaptopropyl)triethoxysilane
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contains : (3-Mercaptopropyl)triethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'