missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11319505
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55289-06-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD02094039
InChI Key JPCISVSOTKMFPG-UHFFFAOYSA-N
Synonym 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid
PubChem CID 7021483
IUPAC Name 3-methoxy-2-methylbenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1C

Specifications

Quantity 10 g
Beilstein 510395
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 3-Methoxy-2-methylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'