Learn More
3'-Methoxyacetophenone, 97%, Thermo Scientific Chemicals
23.92 € - 219.09 €
Chemical Identifiers
CAS | 586-37-8 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.177 |
MDL Number | MFCD00008736 |
InChI Key | BAYUSCHCCGXLAY-UHFFFAOYSA-N |
Synonym | 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy |
PubChem CID | 11460 |
IUPAC Name | 1-(3-methoxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=CC=C1)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439160
|
Thermo Scientific Alfa Aesar
A15604.14 |
25 g |
23.92 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449160
|
Thermo Scientific Alfa Aesar
A15604.22 |
100 g |
57.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459160
|
Thermo Scientific Alfa Aesar
A15604.36 |
500 g |
219.09 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.
Solubility
Fully miscible in water. Soluble in alcohol.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
586-37-8 | |
150.177 | |
BAYUSCHCCGXLAY-UHFFFAOYSA-N | |
11460 | |
CC(=O)C1=CC(=CC=C1)OC |
C9H10O2 | |
MFCD00008736 | |
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy | |
1-(3-methoxyphenyl)ethanone |
Specifications
586-37-8 | |
1.094 | |
>110°C (230°F) | |
1.5425 | |
878552 | |
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy | |
BAYUSCHCCGXLAY-UHFFFAOYSA-N | |
1-(3-methoxyphenyl)ethanone | |
11460 | |
97% |
-7°C | |
239°C to 241°C | |
C9H10O2 | |
MFCD00008736 | |
Light sensitive | |
Fully miscible in water. Soluble in alcohol. | |
CC(=O)C1=CC(=CC=C1)OC | |
150.177 | |
150.18 | |
3'-Methoxyacetophenone |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H332
missing translation for 'einecsNumber' : 209-573-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only