missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11373936
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10130-74-2
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD01318155
InChI Key JHJKSEKUZNJKGO-UHFFFAOYSA-N
Synonym 3-methoxybenzene-1-sulfonyl chloride, 3-methoxy-benzenesulfonyl chloride, 3-methoxybenzenesulphonyl chloride, m-anisolesulfonyl chloride, 3-methoxybenzenesulfonylchloride, benzenesulfonyl chloride, 3-methoxy, akos bb-9459, 3-methoxyphenylsulfonyl chloride, 3-methoxybenzene sulfonyl chloride, m-anisolesulphonyl chloride
PubChem CID 2737504
IUPAC Name 3-methoxybenzenesulfonyl chloride
SMILES COC1=CC(=CC=C1)S(=O)(=O)Cl

Specifications

Density 1.46
Boiling Point 158°C to 159°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.557
Quantity 1 g
UN Number UN3265
Beilstein 2093871
Sensitivity Moisture sensitive
Formula Weight 206.65
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'