Learn More
3-Methoxybenzoyl chloride, 99%, Thermo Scientific Chemicals
42.51 € - 574.86 €
Chemical Identifiers
CAS | 1711-05-3 |
---|---|
Molecular Formula | C8H7ClO2 |
Molecular Weight (g/mol) | 170.592 |
MDL Number | MFCD00000673 |
InChI Key | RUQIUASLAXJZIE-UHFFFAOYSA-N |
Synonym | m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid |
PubChem CID | 74374 |
IUPAC Name | 3-methoxybenzoyl chloride |
SMILES | COC1=CC=CC(=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475597
|
Thermo Scientific Alfa Aesar
B23571.09 |
10 g |
42.51 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485597
|
Thermo Scientific Alfa Aesar
B23571.18 |
50 g |
167.84 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495597
|
Thermo Scientific Alfa Aesar
B23571.30 |
250 g |
574.86 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Methoxybenzoyl chloride is used in the synthesis of 2-arylbenzofuran-based molecules that act against Alzheimer’s disease, combating symptoms. In addition, it aids in the optimization of dihydropyrrolopyrimidine inhibitors against PI3K (phosphoinositide 3-kinase) resulting in tumor growth inhibition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in chloroform. Reacts with water.
Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
Chemical Identifiers
1711-05-3 | |
170.592 | |
RUQIUASLAXJZIE-UHFFFAOYSA-N | |
74374 | |
COC1=CC=CC(=C1)C(=O)Cl |
C8H7ClO2 | |
MFCD00000673 | |
m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid | |
3-methoxybenzoyl chloride |
Specifications
1711-05-3 | |
123°C to 125°C (15 mmHg) | |
C8H7ClO2 | |
MFCD00000673 | |
386659 | |
m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid | |
RUQIUASLAXJZIE-UHFFFAOYSA-N | |
3-methoxybenzoyl chloride | |
74374 | |
99% |
1.214 | |
92°C (197°F) | |
1.5586 | |
UN1729 | |
Moisture sensitive | |
Soluble in chloroform. Reacts with water. | |
COC1=CC=CC(=C1)C(=O)Cl | |
170.592 | |
170.6 | |
3-Methoxybenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
missing translation for 'einecsNumber' : 216-975-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.