missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Product Code. 11489582
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1533748
MDL Number MFCD00004386

Specifications

Melting Point 117°C to 121°C
Molecular Formula C10H10O3
Quantity 100 g
Synonym 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid
InChI Key LZPNXAULYJPXEH-AATRIKPKSA-N
SMILES COC1=CC=CC(=C1)C=CC(=O)O
IUPAC Name (E)-3-(3-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 178.19
PubChem CID 637668
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Methoxycinnamic acid, predominantly trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'