missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxyphenylglyoxal hydrate, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 32025-65-3 |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H8O3 |
MDL Number | MFCD08533286 |
Quantity | 5 g |
Synonym | 3-methoxyphenylglyoxal hydrate, 2-3-methoxyphenyl-2-oxoacetaldehyde hydrate, acmc-1c24s, bestipharma 555-843, 3-methoxyphenylglyoxal monohydrate, ethanone,2,2-dihydroxy-1-3-methylphenyl, 3-methoxyphenyl oxo acetaldehyde-water 1/1, 2-3-methoxyphenyl-2-oxidanylidene-ethanal hydrate |
InChI Key | SIHYQEYAJMDKQH-UHFFFAOYSA-N |
SMILES | COC1=CC=CC(=C1)C(=O)C=O |
IUPAC Name | 2-(3-methoxyphenyl)-2-oxoacetaldehyde;hydrate |
Molecular Weight (g/mol) | 164.16 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction