missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, 98%, pure, Thermo Scientific Chemicals

63.72 € - 1673.33 €

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10255270
View Documents
Thermo Scientific Acros
126485000
500 mL Glass bottle
63.72 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10113560
View Documents
Thermo Scientific Acros
126480010
1 L Glass bottle
82.61 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10703701
View Documents
Thermo Scientific Acros
126480025
2.5 L Glass bottle
170.85 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10236923
View Documents
Thermo Scientific Acros
126480250
25 L Metal Drum
1673.33 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-51-3
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
3-methylbutan-1-ol
C5H12O
MFCD00002934
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CHEBI:15837
CC(C)CCO

Specifications

123-51-3
100.0
0.8000g/mL
45°C
97.5% min. (GC)
(CH3)2CHCH2CH2OH
MFCD00002934
15, 5241
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methylbutan-1-ol
31260
4.3 mPa.s (20°C)
98%
Liquid
97.5
-70.0°C
130.0°C
Authentic
C5H12O
1.4050 to 1.4070
01, 392
0.8
Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,oils,,chloroform
CC(C)CCO
88.15
CHEBI:15837
88.15
Pure
3-Methyl-1-butanol
Videos
Sécurité et traitement

Sécurité et traitement

GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-633-5

Identifiant produit
  • 3-Methyl-1-butanol

Mention d'avertissement
  • Warning

Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

FDS
Documents

Documents

Certificats

RUO – Research Use Only