missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10789901
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38894-11-0
Molecular Formula HCl·H2O
Molecular Weight (g/mol) 233.72
MDL Number MFCD00149370
InChI Key IYXXQOGEFHAQGU-PIHABLKOSA-N
Synonym 2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride
PubChem CID 6508671
IUPAC Name (E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 270.0°C to 276.0°C
Color White
Loss on Drying 8% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Sensitivity (to hydroxyl groups) Passes Test
Solubility Information Solubility in water: soluble
Formula Weight 233.72
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'