missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 270.0°C to 276.0°C |
Color | White |
Loss on Drying | 8% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 250 g |
Sensitivity | (to hydroxyl groups) Passes Test |
Show More |
Safety and Handling
Product Identifier
- 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'