missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals

Product Code. 11497667
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-18-6
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00001581
InChI Key IITQJMYAYSNIMI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
PubChem CID 14511
IUPAC Name 3-methylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CCC1

Specifications

Density 0.971
Boiling Point 199°C to 200°C
Flash Point 68°C (154°F)
Odor Pleasant
Refractive Index 1.494
Quantity 100 g
Beilstein 1560601
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 110.15
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-cyclohexen-1-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'