Learn More
3-Methyl-2-cyclopenten-1-one, 98%, stabilized, Thermo Scientific Chemicals
70.30 € - 1190.00 €
Chemical Identifiers
CAS | 2758-18-1 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.13 |
InChI Key | CHCCBPDEADMNCI-UHFFFAOYSA-N |
Synonym | 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one |
PubChem CID | 17691 |
IUPAC Name | 3-methylcyclopent-2-en-1-one |
SMILES | CC1=CC(=O)CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10285730
|
Thermo Scientific Acros
163590100 |
10 g | Glass bottle |
70.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317870
|
Thermo Scientific Acros
163590500 |
50 g | Glass bottle |
262.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914841
|
Thermo Scientific Acros
163592500 |
250 g | Glass Bottle |
1190.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2758-18-1 | |
96.13 | |
3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one | |
3-methylcyclopent-2-en-1-one |
C6H8O | |
CHCCBPDEADMNCI-UHFFFAOYSA-N | |
17691 | |
CC1=CC(=O)CC1 |
Specifications
2758-18-1 | |
Brown-Yellow to Yellow | |
74.0°C (15.0 mmHg) | |
Authentic | |
C6H8O | |
1.4870 to 1.4900 | |
0.98 | |
Solubility in water: miscible | |
CC1=CC(=O)CC1 | |
0.1% hydroquinone | |
17691 | |
98% | |
3-Methyl-2-cyclopenten-1-one |
3.0°C to 5.0°C | |
0.9800g/mL | |
65°C | |
97.5% min. (GC) | |
CH3C5H5(=O) | |
07,I,46 | |
3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one | |
CHCCBPDEADMNCI-UHFFFAOYSA-N | |
3-methylcyclopent-2-en-1-one | |
96.13 | |
96.13 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-421-5
- 3-Methyl-2-cyclopenten-1-one, stabilized
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only