missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10473612
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 849066-63-3
Molecular Formula C9H8N2O4
Molecular Weight (g/mol) 208.173
MDL Number MFCD01880302
InChI Key ASBPDKJMOPNMLF-UHFFFAOYSA-N
Synonym 3-methyl-5-5-methylisoxazol-3-yl isoxazole-4-carboxylic acid, 3-methyl-5-5-methyl-1,2-oxazol-3-yl-1,2-oxazole-4-carboxylic acid, 3,5'-biisoxazole-4'-carboxylicacid, 3',5-dimethyl, maybridge3_007228, 3',5-dimethyl-3,5'-biisoxazole-4'-carboxylic acid, 3-methyl-5-5-methylisoxazol-3-yl-4-carboxylic acid
PubChem CID 2744621
IUPAC Name 3-methyl-5-(5-methyl-1,2-oxazol-3-yl)-1,2-oxazole-4-carboxylic acid
SMILES CC1=CC(=NO1)C2=C(C(=NO2)C)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.17
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.