missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15475437
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-52-5
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD09025371
InChI Key CCKDSMKGHHIBFA-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethoxy benzoyl chloride, 5-methyl-3-trifluoromethoxy benzoyl chloride
PubChem CID 46737538
IUPAC Name 3-methyl-5-(trifluoromethoxy)benzoyl chloride
SMILES CC1=CC(=CC(=C1)C(=O)Cl)OC(F)(F)F

Specifications

Refractive Index 1.4706
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethoxy)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'