missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Boiling Point | 106°C (30mmHg) |
Quantity | 25 g |
Beilstein | 2072720 |
Flash Point | 82°C (179°F) |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 193.32 |
Percent Purity | 95% |
Refractive Index | 1.419 |
Sensitivity | Moisture sensitive |
Density | 0.978 |
Show More |
Safety and Handling
Product Identifier
- (3-Methylaminopropyl)trimethoxysilane
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : (3-Methylaminopropyl)trimethoxysilane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'