missing translation for 'onlineSavingsMsg'
Learn More

3-Methylanisole, 99%, Thermo Scientific Chemicals

Product Code. 11486964
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-84-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008395
InChI Key OSIGJGFTADMDOB-UHFFFAOYSA-N
Synonym 3-methylanisole, 3-methoxytoluene, m-methylanisole, m-cresol methyl ether, m-methoxytoluene, benzene, 1-methoxy-3-methyl, anisole, m-methyl, methyl m-tolyl ether, m-cresyl methyl ether, methyl m-cresyl ether
PubChem CID 7530
IUPAC Name 1-methoxy-3-methylbenzene
SMILES COC1=CC=CC(C)=C1

Specifications

Density 0.969
Boiling Point 175°C to 176°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.513
Quantity 50 g
UN Number UN3271
Beilstein 907023
Solubility Information Soluble in alcohol and insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 3-Methylanisole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylanisole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : 3-Methylanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'