missing translation for 'onlineSavingsMsg'
Learn More

3-Methylflavone-8-carboxylic acid, 98+%, Thermo Scientific™

Product Code. 11397696
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3468-01-7
MDL Number MFCD00040998

Specifications

Melting Point 234°C to 236°C
Molecular Formula C10H10O
Quantity 5 g
Beilstein 1433509
Synonym cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
InChI Key PJRHFTYXYCVOSJ-UHFFFAOYSA-N
SMILES C1CC1C(=O)C2=CC=CC=C2
IUPAC Name cyclopropyl(phenyl)methanone
Molecular Weight (g/mol) 146.19
PubChem CID 77029
Formula Weight 280.28
Percent Purity ≥98%
Chemical Name or Material 3-Methylflavone-8-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.