missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole, 99%, Thermo Scientific Chemicals

Product Code. 11316706
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

Solubility
Soluble in water, Ether, Alcohols, Benzene, Acetone, Chloroform.

Notes
Light Sensitive. Store in dark away from oxidizing agents. Incompatible with light, strong acids, acid ahydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-34-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005627
InChI Key ZFRKQXVRDFCRJG-UHFFFAOYSA-N
Synonym 3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
PubChem CID 6736
ChEBI CHEBI:9171
IUPAC Name 3-methyl-1H-indole
SMILES CC1=CNC2=CC=CC=C12

Specifications

Melting Point 94°C to 97°C
Boiling Point 265°C to 268°C
Flash Point 132°C (269°F)
Odor Stench
Quantity 5 g
Beilstein 111296
Sensitivity Light sensitive
Merck Index 14,8560
Solubility Information Soluble in water,Ether,Alcohols,Benzene,Acetone,Chloroform.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 3-Methylindole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'