Learn More
3-Methylpiperidine, 97%, Thermo Scientific Chemicals
23.20 € - 54.80 €
Chemical Identifiers
CAS | 626-56-2 |
---|---|
Molecular Formula | C6H13N |
Molecular Weight (g/mol) | 99.177 |
MDL Number | MFCD00005994 |
InChI Key | JEGMWWXJUXDNJN-UHFFFAOYSA-N |
Synonym | 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc |
PubChem CID | 79081 |
IUPAC Name | 3-methylpiperidine |
SMILES | CC1CCCNC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353037
|
Thermo Scientific Alfa Aesar
L13408.14 |
25 g |
23.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363037
|
Thermo Scientific Alfa Aesar
L13408.22 |
100 g |
54.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
Chemical Identifiers
626-56-2 | |
99.177 | |
JEGMWWXJUXDNJN-UHFFFAOYSA-N | |
79081 | |
CC1CCCNC1 |
C6H13N | |
MFCD00005994 | |
3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc | |
3-methylpiperidine |
Specifications
626-56-2 | |
0.845 | |
8°C (46°F) | |
C6H13N | |
MFCD00005994 | |
79807 | |
It is soluble in water. | |
CC1CCCNC1 | |
99.177 | |
99.18 | |
3-Methylpiperidine |
-24°C | |
124°C to 125°C | |
Amine-like | |
1.447 | |
UN2733 | |
3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc | |
JEGMWWXJUXDNJN-UHFFFAOYSA-N | |
3-methylpiperidine | |
79081 | |
97% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 210-953-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only