missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylthio)benzoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 126°C to 132°C |
Quantity | 25 g |
Formula Weight | 168.21 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Methylthio)benzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'