missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 98+%, Thermo Scientific Chemicals

32.46 € - 210.56 €

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405403
View Documents
Thermo Scientific Alfa Aesar
A13394.14
25 g
32.46 €
25g
Please to purchase this item. Need a web account? Register with us today!
11415403
View Documents
Thermo Scientific Alfa Aesar
A13394.22
100 g
90.45 €
100g
Please to purchase this item. Need a web account? Register with us today!
11425403
View Documents
Thermo Scientific Alfa Aesar
A13394.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-44-4
98.163
QENGPZGAWFQWCZ-UHFFFAOYSA-N
12024
CC1=CSC=C1
C5H6S
MFCD00005470
thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
3-methylthiophene

Specifications

616-44-4
1.02
11°C (51°F)
C5H6S
MFCD00005470
1300
QENGPZGAWFQWCZ-UHFFFAOYSA-N
3-methylthiophene
12024
≥98%
-69°C
115°C to 117°C
Aromatic
1.52
UN1993
thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
CC1=CSC=C1
98.163
98.17
3-Methylthiophene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H312-H315-H319-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 210-482-6

missing translation for 'rtecsNumber' : XM9800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.