missing translation for 'onlineSavingsMsg'
Learn More

3-Morpholinobenzoic acid, 97%, Thermo Scientific™

Product Code. 10016202
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 215309-00-5
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD06659078
InChI Key VSKFQESEPGOWBS-UHFFFAOYSA-N
Synonym 3-morpholinobenzoic acid, 3-4-morpholinyl benzoic acid, 3-morpholin-4-yl-benzoic acid, 3-morpholin-4-yl benzoic acid, 3-morpholinobenzoicacid, benzoic acid, 3-4-morpholinyl, 4-3-carboxyphenyl morpholine, pubchem19534, 3-4-morpholino benzoic acid
PubChem CID 2795549
IUPAC Name 3-morpholin-4-ylbenzoic acid
SMILES C1COCCN1C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 3-Morpholinobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.