missing translation for 'onlineSavingsMsg'
Learn More

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Product Code. 10187992
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 145127-38-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD07772812
InChI Key MXBVALXTJZMIJB-UHFFFAOYSA-N
PubChem CID 7162074
IUPAC Name (3-morpholin-4-ylphenyl)methanol
SMILES OCC1=CC(=CC=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (3-Morpholinophenyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.